ZymoPURE II™ Plasmid Maxiprep Kits

Supplier: Zymo Research

D4202 D4203
77001-468EA 349.46 USD
77001-468 77001-470
ZymoPURE II™ Plasmid Maxiprep Kits
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

Part of the ZymoPURE plasmid kits collection, the ZymoPURE II plasmid maxiprep kits provides the fastest and simplest method available to efficiently isolate up to 3 mg of transfection grade plasmid DNA from E. coli.



Utilizing a modified alkaline lysis in conjunction with a binding system, the ZymoPURE IImaxiprep kits can process up to 150 ml of culture in less than 18 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is rapidly bound onto a column with either a vacuum or a centrifuge instead of a slow gravity flow column. Additionally, there is no alcohol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 6 µg/µl), endotoxin-free and transfection-ready. As an added convenience, the ZymoPURE II plasmid maxiprep kit contains colored buffers that permit error-free visualization and identification of complete bacterial cell lysis and neutralization.


Order Now

SPECIFICATIONS

Description ZymoPURE™ II Plasmid Maxiprep Kit
Environmentally Preferable
Application
Target molecule Plasmid
Refine target molecule Plasmid (endotoxin-free)
Sample type Bacteria
Method / Format Column (silica-based)
Prep size Maxi
Sample Size ≤150 ml
Yield ≤1.2 mg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR