(S)-2-Methylazetidine hydrochloride 98%

Supplier: Ambeed
Warning

A977971-1G
76994-080EA 234.46 USD
76994-080
(S)-2-Methylazetidine hydrochloride 98%
(S)-2-Methylazetidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClN
MW: 107.58 g/mol
MDL Number: MFCD29920004
CAS Number: 935669-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR