3-Ethoxy-5-hydroxybenzaldehyde 96%

Supplier: Ambeed
Warning

A253991-250MG
76987-974EA 31.83 USD
76987-974
3-Ethoxy-5-hydroxybenzaldehyde 96%
3-Ethoxy-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.17 g/mol
MDL Number: MFCD20485677
CAS Number: 951800-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR