6-Bromo-4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A105858-250MG
76982-398EA 187.78 USD
76982-398
6-Bromo-4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
6-Bromo-4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrClN
MW: 276.60 g/mol
MDL Number: MFCD16618981
CAS Number: 1203684-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR