4-Bromo-1,2-benzisoxazol-3-amine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromobenzo[d]isoxazol-3-amine

A141911-100MG A141911-1G A141911-250MG A141911-5G
76972-636EA 8 USD
76972-636 76972-638 76972-640 76972-642
4-Bromo-1,2-benzisoxazol-3-amine 98%
4-Bromo-1,2-benzisoxazol-3-amine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MW: 213.0300 g/mol
MDL Number: MFCD11845370
CAS Number: 796969-15-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR