Benzyl 3-hydroxycyclobutanecarboxylate 97%

Supplier: Ambeed
Warning

A112460-250MG
76970-902EA 29.71 USD
76970-902
Benzyl 3-hydroxycyclobutanecarboxylate 97%
Benzyl 3-hydroxycyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.238 g/mol
MDL Number: MFCD11848893
CAS Number: 480449-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR