Anti-Cyclin A2 Mouse Monoclonal Antibody [clone: CCNA2/2333]

Supplier: ANTIBODIES.COM LLC
A250453-100
76968-390EA 555.5 USD
76968-390
Anti-Cyclin A2 Mouse Monoclonal Antibody [clone: CCNA2/2333]
Antibodies
Mouse monoclonal [CCNA2/2333] antibody to Cyclin A2 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cyclin A2
Clonality: Monoclonal
Clone: CCNA2/2333
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR