Anti-Mouse IgG2a Mouse Monoclonal Antibody [Clone: MOPC-173] (APC (Allophycocyanin))

Supplier: ANTIBODIES.COM LLC
A86300-100
76964-186EA 402.6 USD
76964-186
Anti-Mouse IgG2a Mouse Monoclonal Antibody [Clone: MOPC-173] (APC (Allophycocyanin))
Antibodies
Mouse IgG2a [MOPC-173] (APC) - Isotype Control for Flow Cytometry and Isotype Control.

  • Validated Applications: Flow Cytometry, Isotype Control
  • Recommended Dilutions: Isotype Control: Use the same concentration of this isotype control as the working concentration of the primary antibody

Type: Control
Antigen: Mouse IgG2a
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR