Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469 + BRD494 + BRD.3]

Supplier: ANTIBODIES.COM LLC
A250997-100
76964-112EA 555.5 USD
76964-112
Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469 + BRD494 + BRD.3]
Antibodies
Mouse monoclonal [BRD469 + BRD494 + BRD.3] antibody to BrdU for IHC-P.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml
  • Host: Mouse
  • Clonality: Oligoclonal
  • Isotype: IgG1, kappa + IgG1, kappa + IgG1, kappa
  • Conjugate: Unconjugated

Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: BRD469 + BRD494 + BRD.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR