Anti-EPX Mouse Monoclonal Antibody [clone: AHE-1]

Supplier: ANTIBODIES.COM LLC
A253583-100
76961-270EA 555.5 USD
76961-270
Anti-EPX Mouse Monoclonal Antibody [clone: AHE-1]
Antibodies
Mouse monoclonal [AHE-1] antibody to EPX for ELISA with samples derived from Human.

  • Validated Applications: ELISA

Type: Primary
Antigen: EPX
Clonality: Monoclonal
Clone: AHE-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR