Anti-CD130 (gp130) Mouse Monoclonal Antibody [Clone: IL6ST/4101]

Supplier: ANTIBODIES.COM LLC
A277672-100
76953-888EA 555.5 USD
76953-888
Anti-CD130 (gp130) Mouse Monoclonal Antibody [Clone: IL6ST/4101]
Antibodies
Mouse monoclonal [IL6ST/4101] antibody to CD130 (gp130) for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD130 (gp130)
Clonality: Monoclonal
Clone: IL6ST/4101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR