Anti-FOXA1 Mouse Monoclonal Antibody [clone: FOXA1/1241]

Supplier: ANTIBODIES.COM LLC
A252041-100
76952-118EA 555.5 USD
76952-118
Anti-FOXA1 Mouse Monoclonal Antibody [clone: FOXA1/1241]
Antibodies
Mouse monoclonal [FOXA1/1241] antibody to FOXA1 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human and Rat.

  • Validated Applications: Flow Cytometry, IF, WB, IHC-P
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: FOXA1
Clonality: Monoclonal
Clone: FOXA1/1241
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Rat
Order Now

SPECIFICATIONS

Antigen Symbol FOXA1
Antigen Name forkhead box A1
Antigen Synonyms HNF-3-alpha|FOXA1|hepatocyte nuclear factor 3 alpha|FOXA1_HUMAN|FOX A1|Hepatocyte nuclear factor 3-alpha|Forkhead box protein A1|HNF-3A|HNF 3A
Antibody Type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone FOXA1/1241
Reactivity Human,Rat
Host Mouse
Gene ID UniprotID# P55317
Isotype IgG2b kappa
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Immunogen Recombinant fragment, around amino acids 372 - 472, of human FOXA1 protein. The exact sequence is proprietary
Form Liquid
Molecular Weight 49 kDa
Purification Protein A/G chromatography
Concentration 1 mg/ml
Storage Buffer Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
Storage Temperature Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
Shipping Temperature Shipped on blue ice at 4 ℃
Tested Applications IHC-P

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR