PIP® Bouton® Universal Fit Polycarbonate Safety Visor
Supplier: PIP
251-01-5201
76944-632CS 330.6 USD
76944-632
PIP® Bouton® Universal Fit Polycarbonate Safety Visor
Face Shields
Universal fit polycarbonate safety visor with 0.040" thick lens.
  • Made with virgin resin for better uniformity of the viewing area
  • Universal hole pattern will fit most headgear and hard hat adapters
  • Ships with masking on both sides to prevent scratching
Polycarbonate safety visor with aluminum binding is ideal for high impact applications. Outperforms PETG (Polyethylene Terephthalate Glycol) in a wider range of temperatures.
Certificaciones: Meets ANSI Z87.1. Meets CSA Z94.3.