Methyl 5-fluoro-2-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

35464-1G 35464-25G 35464-500MG 35464-5G
76941-208EA 331.01 USD
76941-208 76941-210 76941-212 76941-214
Methyl 5-fluoro-2-(trifluoromethyl)benzoate ≥95%
Methyl 5-fluoro-2-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203755
CAS Number: 773873-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR