4-Formyl-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Formyl-2-methylphenyl)boronic acid

10442-25G
76924-324EA 127.31 USD
76924-324
4-Formyl-2-methylphenylboronic acid ≥97%
4-Formyl-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₃
MW: 163.966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839115
CAS Number: 156428-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR