3-Fluoro-4-(hydrazinocarbonyl)benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-4-hydrazinocarbonylphenylboronic acid

12376-25G 12376-5G
76906-972EA 605.78 USD
76906-972 76906-974
3-Fluoro-4-(hydrazinocarbonyl)benzeneboronic acid ≥97%
3-Fluoro-4-(hydrazinocarbonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFN₂O₃
MW: 197.959 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973743
CAS Number: 850568-06-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR