2-Methyl-5-nitrobiphenyl ≥97%

Supplier: AOBChem USA

Warning

25079-1G 25079-25G 25079-500MG 25079-5G
76899-846EA 266.29 USD
76899-846 76899-848 76899-850 76899-852
2-Methyl-5-nitrobiphenyl ≥97%
2-Methyl-5-nitrobiphenyl
Formula: C₁₃H₁₁NO₂
MW: 213.232 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02179253
CAS Number: 13480-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR