(2-Methoxy-2'-methyl-[1,1'-biphenyl]-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

61539-1G 61539-25G 61539-500MG 61539-5G
76892-504EA 257.8 USD
76892-504 76892-506 76892-508 76892-510
(2-Methoxy-2'-methyl-[1,1'-biphenyl]-4-yl)methanol ≥97%
(2-Methoxy-2'-methyl-[1,1'-biphenyl]-4-yl)methanol
Formula: C₁₅H₁₆O₂
MW: 228.2863 g/mol
Storage Temperature: Ambient
CAS Number: 2918925-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR