3-Chloro-5-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-iodophenyl methyl ether, 1-Chloro-3-iodo-5-methoxybenzene

53107-1G 53107-25G 53107-500MG 53107-5G
76891-354EA 379.81 USD
76891-354 76891-356 76891-358 76891-360
3-Chloro-5-iodoanisole ≥97%
3-Chloro-5-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆ClIO
MW: 268.479 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18393318
CAS Number: 861800-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR