2,4-Dibromo-6-methylphenol ≥97%

Supplier: AOBChem USA

Warning

45558-25G
76889-976EA 144.29 USD
76889-976
2,4-Dibromo-6-methylphenol ≥97%
2,4-Dibromo-6-methylphenol
Formula: C₇H₆Br₂O
Storage Temperature: Ambient
MDL Number: MFCD00152390
CAS Number: 609-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR