2-(Benzyloxy)-4-bromophenol ≥97%

Supplier: AOBChem USA

Warning

32277-25G
76887-420EA 162.32 USD
76887-420
2-(Benzyloxy)-4-bromophenol ≥97%
2-(Benzyloxy)-4-bromophenol

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO₂
MW: 279.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18433365
CAS Number: 153240-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR