tert-Butyl 3-iodobenzoate 96%

Supplier: Ambeed
Warning

A577244-250MG
76878-772EA 40.32 USD
76878-772
tert-Butyl 3-iodobenzoate 96%
tert-Butyl 3-iodobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃IO₂
MW: 304.12 g/mol
MDL Number: MFCD18837617
CAS Number: 173406-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR