5-Ethoxypyrimidin-2-amine 97%

Supplier: Ambeed
Warning

A201283-250MG
76873-408EA 57.29 USD
76873-408
5-Ethoxypyrimidin-2-amine 97%
5-Ethoxypyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MDL Number: MFCD00223638
CAS Number: 39268-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR