4-Bromo-2-chlorophenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromo-2-chlorophenyl)acetic acid

A132797-10G A132797-1G A132797-5G
103801-782EA 29 USD
103801-782 76870-822 77298-580 77314-366
4-Bromo-2-chlorophenylacetic acid 98%
4-Bromo-2-chlorophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MW: 249.489 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847409
CAS Number: 916516-89-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR