Anti-FLVCR2 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A10228-100
76866-580EA 351.7 USD
76866-580 76862-928
Anti-FLVCR2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FLVCR2 for WB with samples derived from Human and Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: FLVCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR