2-Fluoro-5-(trifluoromethyl)pyridine 96%

Supplier: Ambeed
Danger

A631690-5G A631690-10G A631690-1G A631690-25G
103840-274EA 6 USD
103840-274 76844-442 77266-974 77308-012 77308-014
2-Fluoro-5-(trifluoromethyl)pyridine 96%
2-Fluoro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃F₄N
MW: 165.09
Boiling Pt: 135.3 °C (760 mm Hg)
Flash Pt: 32.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07437945
CAS Number: 69045-82-5
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR