Benzophenone imine 97%

Supplier: Ambeed
Warning

Synonyms: Benzophenoneimine

A614769-100G A614769-10G A614769-500G A614769-5G A614769-1000G A614769-25G
103810-482EA 133 USD
103810-482 76696-644 76696-646 76696-648 76696-650 76844-350 76844-352
Benzophenone imine 97%
Benzophenone imine

Precaución: for Research Use Only

Formula: C₁₃H₁₁N
MW: 181.24 g/mol
Boiling Pt: 282 °C (760 mm Hg)
Melting Pt: 30 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00001760
CAS Number: 1013-88-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR