6-Bromo-4-chloropicolinonitrile 95%

Supplier: Ambeed
Warning

A450964-250MG
76829-710EA 87 USD
76829-710
6-Bromo-4-chloropicolinonitrile 95%
6-Bromo-4-chloropicolinonitrile

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂
MW: 217.45 g/mol
MDL Number: MFCD14582023
CAS Number: 1206248-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR