2-Amino-6-bromoquinazolin-4-ol 95%

Supplier: Ambeed
Warning

A284325-100MG A284325-250MG
103813-634EA 139 USD
103813-634 76827-602 76827-604
2-Amino-6-bromoquinazolin-4-ol 95%
2-Amino-6-bromoquinazolin-4-ol

Precaución: for Research Use Only

Formula: C₈H₆BrN₃O
MW: 240.057 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16250251
CAS Number: 130148-53-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR