6-Methylpiperidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-2-piperidinecarboxylic acid

A226021-250MG
103819-218EA 196 USD
103819-218 103850-120 76826-318
6-Methylpiperidine-2-carboxylic acid 95%
6-Methylpiperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.18 g/mol
Boiling Pt: 267 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00513436
CAS Number: 99571-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR