1-(2-Amino-5-methylthiazol-4-yl)ethanone 97%

Supplier: Ambeed
Warning

A150987-100MG A150987-1G
76823-816EA 10.61 USD
76823-816 76823-818
1-(2-Amino-5-methylthiazol-4-yl)ethanone 97%
1-(2-Amino-5-methylthiazol-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₈N₂OS
MW: 156.20 g/mol
MDL Number: MFCD08692000
CAS Number: 40353-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR