2,3-Dihydrofuro[2,3-b]pyridin-3-ol 95%

Supplier: Ambeed
Warning

A537668-50MG
76818-964EA 170.81 USD
76818-964
2,3-Dihydrofuro[2,3-b]pyridin-3-ol 95%
2,3-Dihydrofuro[2,3-b]pyridin-3-ol

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.13 g/mol
MDL Number: MFCD18836698
CAS Number: 144186-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR