Ferrous gluconate 98%

Supplier: Ambeed
Warning

Synonyms: Iron(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate

A776067-100G A776067-5G A776067-1000G A776067-25G A776067-500G
76807-352EA 73 USD
76807-352 76807-354 77602-994 77670-078 77670-080
Ferrous gluconate 98%
Ferrous gluconate

Precaución: for Research Use Only

Formula: C₁₂H₂₂FeO₁₄
MW: 446.14 g/mol
MDL Number: MFCD04037236
CAS Number: 299-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR