5-(3-Chlorophenyl)-2-furaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-(3-Chlorophenyl)furan-2-carbaldehyde

A637181-100G A637181-1G A637181-25G A637181-5G
76806-272EA 318 USD
76806-272 76806-274 76806-276 76806-278
5-(3-Chlorophenyl)-2-furaldehyde 98%
5-(3-Chlorophenyl)-2-furaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₇ClO₂
MW: 206.63 g/mol
MDL Number: MFCD00214773
CAS Number: 22078-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR