tert-Butyl-5-aminoisoindoline-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-aminoisoindoline-2-carboxylate

A129361-100MG
103801-272EA 84 USD
103801-272 76782-228
tert-Butyl-5-aminoisoindoline-2-carboxylate 95%
tert-Butyl-5-aminoisoindoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.2942 g/mol
Boiling Pt: 376.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08234717
CAS Number: 264916-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR