Hei-PLATE Mix 'N' Heat Ultimate Magnetic Hotplate Stirrers
Supplier: Heidolph NA, LLC
036110228 036110229 036110232 036110233 036110238 036110239
76777-162EA 2812 USD
76777-162 76777-164 76777-170 76777-172 76777-182 76777-184
Hei-PLATE Mix 'N' Heat Ultimate Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
For smooth to intense mixing and heating of low-viscosity fluids the Hei-PLATE Mix 'N' Heat Ultimate magnetic hotplate stirrers, are an excellent choice for use with decomposing organic and inorganic substances.
  • One display for each main function (stirring and heating) – provides clear view of all values and settings at any time
  • Set parameters via 2 push-and-rotate controls with LED-rings for easy identification of active functions
  • Separate timers for heating and stirring functions mean flexibility and improved control of experiments
  • Choice of 3 heating
More Product Information