4',5'-Dimethoxy-2'-hydroxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-4,5-dimethoxyacetophenone, 1-(2-Hydroxy-4,5-dimethoxyphenyl)ethan-1-one

A533356-1G A533356-250MG A533356-5G
76774-724EA 30 USD
76774-724 76774-726 76774-728
4',5'-Dimethoxy-2'-hydroxyacetophenone 98%
4',5'-Dimethoxy-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MW: 196.1999 g/mol
MDL Number: MFCD00017241
CAS Number: 20628-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR