3,3',5'-Triiodo-L-thyronine sodium salt 95%

Supplier: Ambeed
Warning

Synonyms: Liothyronine sodium salt, Sodium (S)-2-amino-3-(4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl)propanoate

A103275-1G A103275-1MG A103275-250MG A103275-25MG A103275-100MG A103275-5G
76765-302EA 71 USD
76765-302 76765-304 76765-306 76765-308 77623-848 77623-850
3,3',5'-Triiodo-L-thyronine sodium salt 95%
3,3',5'-Triiodo-L-thyronine sodium salt

Precaución: for Research Use Only

Formula: C₁₅H₁₁I₃NNaO₄
MW: 672.959 g/mol
MDL Number: MFCD00002594
CAS Number: 55-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR