Methyl-4-methyl-4-nitropentanoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-methyl-4-nitropentanoate

A743975-100MG A743975-1G A743975-250MG A743975-5G
76728-094EA 18 USD
76728-094 76728-096 76728-098 77670-010
Methyl-4-methyl-4-nitropentanoate 98%
Methyl-4-methyl-4-nitropentanoate

Precaución: for Research Use Only

Formula: C₇H₁₃NO₄
MW: 175.19 g/mol
MDL Number: MFCD00047757
CAS Number: 16507-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR