N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic acid) 97%

Supplier: Ambeed
Danger

Synonyms: ((Ethane-1,2-diylbis(azanetriyl))tetrakis(methylene))tetraphosphonic acid

A662547-25G A662547-5G
76727-452EA 20 USD
76727-452 76727-454
N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic acid) 97%
N,N,N',N'-Ethylenediaminetetrakis(methylenephosphonic acid)

Precaución: for Research Use Only

Formula: C₆H₂₀N₂O₁₂P₄
MW: 436.12 g/mol
MDL Number: MFCD00058893
CAS Number: 1429-50-1
UN: 1759
ADR: 8 Ⅲ

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR