1-(2-Chlorophenyl)urea 95%

Supplier: Ambeed
Warning

A495137-100MG A495137-1G A495137-25G
76725-984EA 7 USD
76725-984 76725-986 76725-988
1-(2-Chlorophenyl)urea 95%
1-(2-Chlorophenyl)urea

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
MW: 170.59 g/mol
MDL Number: MFCD00041316
CAS Number: 114-38-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR