N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-(Cyclohexyloxy)-4-nitrophenyl)methanesulphonamide, NS-398, N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide

A113156-10MG A113156-1MG A113156-50MG A113156-5MG
76718-200EA 23 USD
76718-200 76718-202 76718-204 76718-206
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulfonamide 98%
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulfonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₅S
MW: 314.356 g/mol
MDL Number: MFCD00882995
CAS Number: 123653-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR