AccuForge® GEM® Single Edge Blade Cartridges, Uncoated
Supplier: Accutec Blades
AGBL-7009-0000 AGBL-7010-0000 AGBL-7014-BULK
76717-302CS 984.37 USD
76717-302 76717-304 76717-306
AccuForge® GEM® Single Edge Blade Cartridges, Uncoated
Razors and Blades
AccuForge single edge blades feature 3-facet edges for sharpness. Blades are degreased to remove residual oil.
  • 3-facet edge for increased sharpness
  • Degreased to remove oil residue
  • Paper-free packaging is clean room ready
  • Individually packed in clamshell with product key and instructions for use
  • Available in bulk pack without clamshells
  • Each cartridge contains 100 blades
Información para pedidos: For use with GEM RUNNER™ Dispenser Cat no. 76717-342.
Precaución: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending bladesMore Product Information