Sapphire FL Biomolecular Imager
Supplier: Azure Biosystems
IS4000 IS4001 IS4002 IS4003 IS4004 IS4005 IS4006 IS4007 IS4059 IS4027 IS4055 IS4056 IS4057 IS4058
76716-988EA 82092.54 USD
76716-988 76716-950 76716-954 76716-996 76716-956 76717-006 76716-992 76716-952 77449-146 77449-126 77449-138 77449-140 77449-142 77449-144
Sapphire FL Biomolecular Imager
Photodocumentation Systems
Flexibility with uncompromising performance – from in vitro molecular assays to in vivo imaging.
  • Sapphire FL is the ultimate biomolecular imager for flexibility
  • Laser scanning system designed for western blots, 96 well plates, animal imaging and more
With customizable and user-changeable laser and filter modules, the Sapphire FL easily adapts to a lab’s changing needs and advancing research. The Sapphire FL offers customizable and user-changeable optical modules, 5 to 1000 µm resolution scans, a Z-plane range from −1.0 to +6 mm, 5 anesthesia ports forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable