Lighthouse Chemo Bio-Wipe Bag
Supplier: Unimed-Midwest
MLBM015111C
76716-820BX 157.95 USD
76716-820
Lighthouse Chemo Bio-Wipe Bag
Bags Waste Disposal Bags
Quick, economical cleanup for biohazardous Chemo spills.
  • Provides a convenient, temporary containment vehicle for proper disposal into an approved OSHA/ASHP container
  • Reduces risk of contact with harmful chemotherapy drugs, when used with chemo-tested medical exam gloves
  • Complies with OSHA Standard 29CFR 1910.1030 for PPE
  • Composed of a patented chemo yellow opaque polyethylene plastic bag on one side, and an absorbable pad bonded to the other side
The absorbent pad (11.75×9") capacity is approximately 1/2 pint. An adhesive strip provides closure forMore Product Information