Mouse Retinol-binding Protein,RBP ELISA Kit

Supplier: AFG BIOSCIENCE LLC

EK731543
76713-556EA 380.58 USD
76713-556
Mouse Retinol-binding Protein,RBP ELISA Kit
Assays ELISAs

  • Detection wavelength: 450 nm

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein Retinol-binding protein,RBP
Primary antibody reactivity
Assay Type (ELISA with LOV) Sandwich
Host Mouse
Sensitivity 1 - 5 h
Sample Type Serum, plasma, urine, tissue homogenates, cell culture supernates
Sample Volume 50 - 100 μl
Time to Results 1 - 5 h
Regulatory Status RUO
Storage Temperature 2…8 °C
Shelf Life 6 months (4 °C); 12 months (−20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR