Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane) 98%

Supplier: Ambeed
Warning

Synonyms: 6,7,9,10,12,13,15,16,23,24,26,27,29,30,32,33-Hexadecahydrodibenzo[b,q][1,4,7,10,13,16,19,22,25,28]decaoxacyclotriacontine, Dibenzo-30-crown-10-ether

A829731-100MG A829731-1G A829731-250MG
76698-234EA 45 USD
76698-234 76698-236 76698-238
Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane) 98%
Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane)

Precaución: for Research Use Only

Formula: C₂₈H₄₀O₁₀
MW: 536.62 g/mol
MDL Number: MFCD00054624
CAS Number: 17455-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR