3,5-Dimethylphenyl isocyanate 98%

Supplier: Ambeed
Danger

Synonyms: 3,5-Xylyl isocyanate, 3,5-Dimethylphenylisocyanate

A792944-1G A792944-5G A792944-25G
76697-972EA 10 USD
76697-972 76697-974 77627-582
3,5-Dimethylphenyl isocyanate 98%
3,5-Dimethylphenyl isocyanate

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 205
MDL Number: MFCD00013868
CAS Number: 54132-75-1
UN: 2206
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR