Morpholino benzoate 95%

Supplier: Ambeed
Warning

A704856-100MG A704856-1G A704856-250MG A704856-5G
76697-342EA 26 USD
76697-342 76697-344 76697-346 76697-348
Morpholino benzoate 95%
Morpholino benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MW: 207.22 g/mol
MDL Number: MFCD09032166
CAS Number: 5765-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR