Norbornane-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Bicyclo[2.2.1]heptane-2-carboxylic acid

A655635-100G A655635-10G A655635-1G A655635-25G A655635-5G
76697-000EA 416 USD
76697-000 76697-002 76697-004 76697-006 76697-008
Norbornane-2-carboxylic acid 98%
Norbornane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MW: 140.18 g/mol
MDL Number: MFCD00167749
CAS Number: 824-62-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR